2-(5-Oxazolyl)benzylamine
Lieferant: Apollo Scientific
OR470183-1G
OR470183-5G
APOSOR470183-1GEA
397
EUR
APOSOR470183-1G
APOSOR470183-5G
2-(5-Oxazolyl)benzylamine
2-(5-Oxazolyl)benzylamine
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174.199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375280 CAS-Nummer: 1261268-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...