2-(4-Nitrophenyl)imidazole

Lieferant: Apollo Scientific

Synonyme: 2-(4-Nitrophenyl)-1H-imidazole

OR470195-1G OR470195-5G
APOSOR470195-1GEA 306 EUR
APOSOR470195-1G APOSOR470195-5G
2-(4-Nitrophenyl)imidazole
2-(4-Nitrophenyl)imidazole
Formel: C₉H₇N₃O₂
Molecular Weight: 189,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08668825
CAS-Nummer: 1614-06-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR