5-Amino-3-(2-ethoxyphenyl)isoxazole

Lieferant: Apollo Scientific

OR470201-1G OR470201-5G
APOSOR470201-1GEA 306 EUR
APOSOR470201-1G APOSOR470201-5G
5-Amino-3-(2-ethoxyphenyl)isoxazole
5-Amino-3-(2-ethoxyphenyl)isoxazole
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07383033
CAS-Nummer: 501902-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR