(S)-2-(Hydroxymethyl)-N,N-dimethylpyrrolidine-1-carboxamide

Lieferant: Apollo Scientific

OR470214-1G OR470214-5G
APOSOR470214-1GEA 306 EUR
APOSOR470214-1G APOSOR470214-5G
(S)-2-(Hydroxymethyl)-N,N-dimethylpyrrolidine-1-carboxamide
(S)-2-(Hydroxymethyl)-N,N-dimethylpyrrolidine-1-carboxamide
Formel: C₈H₁₆N₂O₂
Molecular Weight: 172.225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651718
CAS-Nummer: 376629-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR