(R)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid

Lieferant: Apollo Scientific

OR470216-1G OR470216-5G
APOSOR470216-1GEA 306 EUR
APOSOR470216-1G APOSOR470216-5G
(R)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid
(R)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid
Formel: C₈H₁₄N₂O₃
Molecular Weight: 186,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17271751
CAS-Nummer: 1046139-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR