1-Isopropyl-2-oxopyrrolidine-4-carboxamide

Lieferant: Apollo Scientific

OR470219-1G OR470219-5G
APOSOR470219-1GEA 256 EUR
APOSOR470219-1G APOSOR470219-5G
1-Isopropyl-2-oxopyrrolidine-4-carboxamide
1-Isopropyl-2-oxopyrrolidine-4-carboxamide
Formel: C₈H₁₄N₂O₂
Molecular Weight: 170,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053006
CAS-Nummer: 90152-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR