1-Boc-5-bromo-6-methyl-1H-indazole

Lieferant: Apollo Scientific

OR470256-1G OR470256-5G
APOSOR470256-1GEA 397 EUR
APOSOR470256-1G APOSOR470256-5G
1-Boc-5-bromo-6-methyl-1H-indazole
1-Boc-5-bromo-6-methyl-1H-indazole
Formel: C₁₃H₁₅BrN₂O₂
Molecular Weight: 311.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16628189
CAS-Nummer: 1305320-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR