1-(2-Pyridyl)-1-butylamine
Lieferant: Apollo Scientific
OR470276-1G
OR470276-5G
APOSOR470276-1GEA
256
EUR
APOSOR470276-1G
APOSOR470276-5G
1-(2-Pyridyl)-1-butylamine
1-(2-Pyridyl)-1-butylamine
Formel:
C₉H₁₄N₂ Molecular Weight: 150.221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08705794 CAS-Nummer: 90565-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...