2-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid

Lieferant: Apollo Scientific

OR470305-5G OR470305-1G
APOSOR470305-5GEA 734 EUR
APOSOR470305-5G APOSOR470305-1G
2-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid
2-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219.237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09046964
CAS-Nummer: 926249-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR