4-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid

Lieferant: Apollo Scientific

OR470314-1G OR470314-5G
APOSOR470314-1GEA 245 EUR
APOSOR470314-1G APOSOR470314-5G
4-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid
4-[(Cyclopropylcarbonyl)amino]-3-methylbenzoic acid
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219.237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09045384
CAS-Nummer: 926224-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR