1-(Boc-amino)-3-bromo-1-(4-tetrahydropyranyl)propane
Lieferant: Apollo Scientific
OR470334-1G
OR470334-5G
APOSOR470334-1GEA
611
EUR
APOSOR470334-1G
APOSOR470334-5G
1-(Boc-amino)-3-bromo-1-(4-tetrahydropyranyl)propane
1-(Boc-amino)-3-bromo-1-(4-tetrahydropyranyl)propane
Formel:
C₁₃H₂₄BrNO₃ Molecular Weight: 322.239 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410600 CAS-Nummer: 924817-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...