Ethyl 6-Phenylpiperidine-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-phenylpiperidine-2-carboxylate

OR470369-1G OR470369-5G
APOSOR470369-1GEA 713 EUR
APOSOR470369-1G APOSOR470369-5G
Ethyl 6-Phenylpiperidine-2-carboxylate
Ethyl 6-Phenylpiperidine-2-carboxylate
Formel: C₁₄H₁₉NO₂
Molecular Weight: 233.306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410519
CAS-Nummer: 1137664-24-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR