6-(2-Methoxyphenyl)-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

OR470370-1G OR470370-5G
APOSOR470370-1GEA 448 EUR
APOSOR470370-1G APOSOR470370-5G
6-(2-Methoxyphenyl)-3-pyridinecarbaldehyde
6-(2-Methoxyphenyl)-3-pyridinecarbaldehyde
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115431
CAS-Nummer: 898405-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR