2-(Fmoc-amino)-2-(4-tetrahydrothiopyranyl)acetic acid

Lieferant: Apollo Scientific

OR470372-1G OR470372-5G
APOSOR470372-1GEA 561 EUR
APOSOR470372-1G APOSOR470372-5G
2-(Fmoc-amino)-2-(4-tetrahydrothiopyranyl)acetic acid
2-(Fmoc-amino)-2-(4-tetrahydrothiopyranyl)acetic acid
Formel: C₂₂H₂₃NO₄S
Molecular Weight: 397,487 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683144
CAS-Nummer: 443991-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR