3-(Boc-amino)-3-(4-chlorophenyl)-1-propanol

Lieferant: Apollo Scientific

OR470388-1G OR470388-5G
APOSOR470388-1GEA 397 EUR
APOSOR470388-1G APOSOR470388-5G
3-(Boc-amino)-3-(4-chlorophenyl)-1-propanol
3-(Boc-amino)-3-(4-chlorophenyl)-1-propanol
Formel: C₁₄H₂₀ClNO₃
Molecular Weight: 285,767 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115553
CAS-Nummer: 886493-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR