[5-(2-Chlorophenyl)-3-pyridyl]methanol
Lieferant: Apollo Scientific
OR470391-1G
OR470391-5G
APOSOR470391-1GEA
499
EUR
APOSOR470391-1G
APOSOR470391-5G
[5-(2-Chlorophenyl)-3-pyridyl]methanol
[5-(2-Chlorophenyl)-3-pyridyl]methanol
Formel:
C₁₂H₁₀ClNO Molecular Weight: 219,667 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410306 CAS-Nummer: 887974-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...