6-(3,4-Dimethoxyphenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470402-1G OR470402-5G
APOSOR470402-1GEA 499 EUR
APOSOR470402-1G APOSOR470402-5G
6-(3,4-Dimethoxyphenyl)nicotinic acid
6-(3,4-Dimethoxyphenyl)nicotinic acid
Formel: C₁₄H₁₃NO₄
Molecular Weight: 259,257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410425
CAS-Nummer: 887976-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR