Dimethyl 2-Acetyl-1,2-dihydroisoquinoline-3,3(4H)-dicarboxylate

Lieferant: Apollo Scientific

OR470419-1G OR470419-5G
APOSOR470419-1GEA 256 EUR
APOSOR470419-1G APOSOR470419-5G
Dimethyl 2-Acetyl-1,2-dihydroisoquinoline-3,3(4H)-dicarboxylate
Dimethyl 2-Acetyl-1,2-dihydroisoquinoline-3,3(4H)-dicarboxylate
Formel: C₁₅H₁₇NO₅
Molecular Weight: 291,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491409
CAS-Nummer: 143767-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR