Ethyl 4-Boc-2-homomorpholinecarboxylate

Lieferant: Apollo Scientific

OR470451-1G OR470451-250MG OR470451-5G
APOSOR470451-1GEA 713 EUR
APOSOR470451-1G APOSOR470451-250MG APOSOR470451-5G
Ethyl 4-Boc-2-homomorpholinecarboxylate
Ethyl 4-Boc-2-homomorpholinecarboxylate
Formel: C₁₃H₂₃NO₅
Molecular Weight: 273,325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21364423
CAS-Nummer: 1363166-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR