6-Methoxyquinazoline-2,4(1H,3H)-dione

Lieferant: Apollo Scientific

OR470456-1G OR470456-5G
APOSOR470456-1GEA 194 EUR
APOSOR470456-1G APOSOR470456-5G
6-Methoxyquinazoline-2,4(1H,3H)-dione
6-Methoxyquinazoline-2,4(1H,3H)-dione
Formel: C₉H₈N₂O₃
Molecular Weight: 192,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954838
CAS-Nummer: 32618-84-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR