2-Methyloxazolo[5,4-b]pyridine
Lieferant: Apollo Scientific
OR470464-1G
OR470464-250MG
APOSOR470464-1GEA
1010
EUR
APOSOR470464-1G
APOSOR470464-250MG
2-Methyloxazolo[5,4-b]pyridine
2-Methyloxazolo[5,4-b]pyridine
Formel:
C₇H₆N₂O Molecular Weight: 134,135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13175745 CAS-Nummer: 91813-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...