6-Hydroxybenzofuran-2-carboxylic acid

Lieferant: Apollo Scientific

OR470478-1G OR470478-250MG
APOSOR470478-1GEA 411 EUR
APOSOR470478-1G APOSOR470478-250MG
6-Hydroxybenzofuran-2-carboxylic acid
6-Hydroxybenzofuran-2-carboxylic acid
Formel: C₉H₆O₄
Molecular Weight: 178,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11101030
CAS-Nummer: 334022-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR