Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride

Lieferant: Apollo Scientific

OR470509-1G OR470509-5G
APOSOR470509-1GEA 397 EUR
APOSOR470509-1G APOSOR470509-5G
Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride
Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride
Formel: C₇H₁₄N₂O₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09998913
CAS-Nummer: 161315-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR