3-(Boc-aminomethyl)pyrazole

Lieferant: Apollo Scientific

OR470562-1G OR470562-250MG
APOSOR470562-1GEA 1330 EUR
APOSOR470562-1G APOSOR470562-250MG
3-(Boc-aminomethyl)pyrazole
3-(Boc-aminomethyl)pyrazole
Formel: C₉H₁₅N₃O₂
Molecular Weight: 197,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14617089
CAS-Nummer: 1251033-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR