N-Methyl-N-[4-(trimethylsilyl)phenyl]pivalamide
Lieferant: Apollo Scientific
OR470571-1G
OR470571-5G
APOSOR470571-1GEA
205
EUR
APOSOR470571-1G
APOSOR470571-5G
N-Methyl-N-[4-(trimethylsilyl)phenyl]pivalamide
N-Methyl-N-[4-(trimethylsilyl)phenyl]pivalamide
Formel:
C₁₅H₂₅NOSi Molecular Weight: 263.451 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22682786 CAS-Nummer: 1418117-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...