Methyl 4-(Cbz-amino)tetrahydropyran-4-carboxylate

Lieferant: Apollo Scientific

OR470606-1G OR470606-5G
APOSOR470606-1GEA 448 EUR
APOSOR470606-1G APOSOR470606-5G
Methyl 4-(Cbz-amino)tetrahydropyran-4-carboxylate
Methyl 4-(Cbz-amino)tetrahydropyran-4-carboxylate
Formel: C₁₅H₁₉NO₅
Molecular Weight: 293.315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23379893
CAS-Nummer: 1427475-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR