Dimethyl 4-Bromo-2-nitrobenzylphosphonate

Lieferant: Apollo Scientific

OR470641-1G OR470641-5G
APOSOR470641-1GEA 397 EUR
APOSOR470641-1G APOSOR470641-5G
Dimethyl 4-Bromo-2-nitrobenzylphosphonate
Dimethyl 4-Bromo-2-nitrobenzylphosphonate
Formel: C₉H₁₁BrNO₅P
Molecular Weight: 324,065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24335258
CAS-Nummer: 1094851-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR