5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole hydrochloride

Lieferant: Apollo Scientific

OR470645-1G OR470645-250MG OR470645-5G
APOSOR470645-1GEA 651 EUR
APOSOR470645-1G APOSOR470645-250MG APOSOR470645-5G
5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole hydrochloride
5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole hydrochloride
Formel: C₅H₆N₂S·ClH
Molecular Weight: 162.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24335251
CAS-Nummer: 1434128-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR