2-Amino-5-bromo-4-(hydroxymethyl)pyridine

Lieferant: Apollo Scientific

OR470672-1G OR470672-250MG
APOSOR470672-1GEA 519 EUR
APOSOR470672-1G APOSOR470672-250MG
2-Amino-5-bromo-4-(hydroxymethyl)pyridine
2-Amino-5-bromo-4-(hydroxymethyl)pyridine
Formel: C₆H₇BrN₂O
Molecular Weight: 203,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13178296
CAS-Nummer: 1227586-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR