Ethyl 3-Bromopyridine-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-bromopicolinate

OR470685-5G OR470685-25G
APOSOR470685-5GEA 75.4 EUR
APOSOR470685-5G APOSOR470685-25G
Ethyl 3-Bromopyridine-2-carboxylate
Ethyl 3-Bromopyridine-2-carboxylate
Formel: C₈H₈BrNO₂
Molecular Weight: 230.059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189685
CAS-Nummer: 434319-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR