3-Hydrazinyl-2-methylpyridine dihydrochloride

Lieferant: Apollo Scientific

OR470719-5G OR470719-1G
APOSOR470719-5GEA 1020 EUR
APOSOR470719-5G APOSOR470719-1G
3-Hydrazinyl-2-methylpyridine dihydrochloride
3-Hydrazinyl-2-methylpyridine dihydrochloride
Formel: C₆H₉N₃·2ClH
Molecular Weight: 196.078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26937550
CAS-Nummer: 1803601-87-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR