5-Bromo-7-chlorobenzofuran

Lieferant: Apollo Scientific

OR470778-1G OR470778-250MG
APOSOR470778-1GEA 1010 EUR
APOSOR470778-1G APOSOR470778-250MG
5-Bromo-7-chlorobenzofuran
5-Bromo-7-chlorobenzofuran
Formel: C₈H₄BrClO
Molecular Weight: 231,474 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11877832
CAS-Nummer: 1194375-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR