2,7-Naphthalenedicarboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: Naphthalene-2,7-dicarboxylic acid

OR470782-1G OR470782-250MG
APOSOR470782-1GEA 938 EUR
APOSOR470782-1G APOSOR470782-250MG
2,7-Naphthalenedicarboxylic acid 97%
2,7-Naphthalenedicarboxylic acid
Formel: C₁₂H₈O₄
Molecular Weight: 216,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060862
CAS-Nummer: 2089-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR