2-[3-(tert-Butoxy)-4-ethoxyphenyl]acetic acid

Lieferant: Apollo Scientific

OR470790-1G OR470790-5G
APOSOR470790-1GEA 397 EUR
APOSOR470790-1G APOSOR470790-5G
2-[3-(tert-Butoxy)-4-ethoxyphenyl]acetic acid
2-[3-(tert-Butoxy)-4-ethoxyphenyl]acetic acid
Formel: C₁₄H₂₀O₄
Molecular Weight: 252,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27500648
CAS-Nummer: 2006277-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR