6-Hydroxy-3-methylbenzo[d]isoxazole-7-carbaldehyde

Lieferant: Apollo Scientific

OR470804-250MG OR470804-1G
APOSOR470804-250MGEA 448 EUR
APOSOR470804-250MG APOSOR470804-1G
6-Hydroxy-3-methylbenzo[d]isoxazole-7-carbaldehyde
6-Hydroxy-3-methylbenzo[d]isoxazole-7-carbaldehyde
Formel: C₉H₇NO₃
Molecular Weight: 177,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386967
CAS-Nummer: 66033-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR