2-Amino-1-isopropylbenzimidazole

Lieferant: Apollo Scientific

OR470830-1G OR470830-250MG OR470830-5G
APOSOR470830-1GEA 142 EUR
APOSOR470830-1G APOSOR470830-250MG APOSOR470830-5G
2-Amino-1-isopropylbenzimidazole
2-Amino-1-isopropylbenzimidazole
Formel: C₁₀H₁₃N₃
Molecular Weight: 175.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159972
CAS-Nummer: 90871-47-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR