2-Benzyl-2,7-diazaspiro[4.5]decane

Lieferant: Apollo Scientific

OR470851-1G OR470851-250MG
APOSOR470851-1GEA 2020 EUR
APOSOR470851-1G APOSOR470851-250MG
2-Benzyl-2,7-diazaspiro[4.5]decane
2-Benzyl-2,7-diazaspiro[4.5]decane
Formel: C₁₅H₂₂N₂
Molecular Weight: 230,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11223500
CAS-Nummer: 1086395-71-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR