Methyl 1-Boc-3-oxopiperidine-2-carboxylate

Lieferant: Apollo Scientific

OR470911-1G OR470911-250MG
APOSOR470911-1GEA 1730 EUR
APOSOR470911-1G APOSOR470911-250MG
Methyl 1-Boc-3-oxopiperidine-2-carboxylate
Methyl 1-Boc-3-oxopiperidine-2-carboxylate
Formel: C₁₂H₁₉NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD15071869
CAS-Nummer: 122019-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR