3-(Boc-amino)-2-butanol

Lieferant: Apollo Scientific

OR470923-1G OR470923-250MG
APOSOR470923-1GEA 1940 EUR
APOSOR470923-1G APOSOR470923-250MG
3-(Boc-amino)-2-butanol
3-(Boc-amino)-2-butanol
Formel: C₉H₁₉NO₃
Molecular Weight: 189,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20232888
CAS-Nummer: 752135-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR