6-Bromo-2-chlorobenzoxazole

Lieferant: Apollo Scientific

OR470937-1G OR470937-250MG OR470937-5G
APOSOR470937-1GEA 601 EUR
APOSOR470937-1G APOSOR470937-250MG APOSOR470937-5G
6-Bromo-2-chlorobenzoxazole
6-Bromo-2-chlorobenzoxazole
Formel: C₇H₃BrClNO
Molecular Weight: 232,462 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11111723
CAS-Nummer: 1260643-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR