2-Methyl-3-(methylsulphonyl)phenylboronic acid

Lieferant: Apollo Scientific

OR470944-1G OR470944-250MG
APOSOR470944-1GEA 2020 EUR
APOSOR470944-1G APOSOR470944-250MG
2-Methyl-3-(methylsulphonyl)phenylboronic acid
2-Methyl-3-(methylsulphonyl)phenylboronic acid
Formel: C₈H₁₁BO₄S
Molecular Weight: 214,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28009326
CAS-Nummer: 1313617-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR