Ethyl 2-(Ethylsulphonyl)acetate

Lieferant: Apollo Scientific

OR470953-1G OR470953-5G
APOSOR470953-1GEA 306 EUR
APOSOR470953-1G APOSOR470953-5G
Ethyl 2-(Ethylsulphonyl)acetate
Ethyl 2-(Ethylsulphonyl)acetate
Formel: C₆H₁₂O₄S
Molecular Weight: 180,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12171498
CAS-Nummer: 29771-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR