2-(1-Pyrazolyl)benzyl alcohol
Lieferant: Apollo Scientific
Synonyme:
2-(1-Pyrazolyl)benzyl Alcohol
OR470991-1G
OR470991-5G
APOSOR470991-1GEA
104
EUR
APOSOR470991-1G
APOSOR470991-5G
2-(1-Pyrazolyl)benzyl alcohol
2-(1-Pyrazolyl)benzyl alcohol
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086188 CAS-Nummer: 741717-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...