3-(5-Oxazolyl)benzaldehyde

Lieferant: Apollo Scientific

OR470992-1G OR470992-5G
APOSOR470992-1GEA 256 EUR
APOSOR470992-1G APOSOR470992-5G
3-(5-Oxazolyl)benzaldehyde
3-(5-Oxazolyl)benzaldehyde
Formel: C₁₀H₇NO₂
Molecular Weight: 173,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24686783
CAS-Nummer: 1097199-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR