5-(Benzyloxy)indolin-2-one

Lieferant: Apollo Scientific

OR471043-1G OR471043-250MG
APOSOR471043-1GEA 916 EUR
APOSOR471043-1G APOSOR471043-250MG
5-(Benzyloxy)indolin-2-one
5-(Benzyloxy)indolin-2-one
Formel: C₁₅H₁₃NO₂
Molecular Weight: 239,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15525665
CAS-Nummer: 156232-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR