3-Chloro-4'-hydroxypropiophenone
Lieferant: Apollo Scientific
OR471082-5G
OR471082-1G
APOSOR471082-5GEA
220
EUR
APOSOR471082-5G
APOSOR471082-1G
3-Chloro-4'-hydroxypropiophenone
3-Chloro-4'-hydroxypropiophenone
Formel:
C₉H₉ClO₂ Molecular Weight: 184.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393259 CAS-Nummer: 7182-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...