2-(6-Hydroxy-1,2-benzisoxazol-3-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

OR471090-1G OR471090-250MG OR471090-5G
APOSOR471090-1GEA 601 EUR
APOSOR471090-1G APOSOR471090-250MG APOSOR471090-5G
2-(6-Hydroxy-1,2-benzisoxazol-3-yl)acetic acid
2-(6-Hydroxy-1,2-benzisoxazol-3-yl)acetic acid
Formel: C₉H₇NO₄
Molecular Weight: 193,156 g/mol
Schmelzpunkt: 186…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20642581
CAS-Nummer: 34173-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR