3-Aminoindole hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 1h-Indol-3-amine hydrochloride

OR471108-1G OR471108-250MG
APOSOR471108-1GEA 543 EUR
APOSOR471108-1G APOSOR471108-250MG
3-Aminoindole hydrochloride 95%
3-Aminoindole hydrochloride
Formel: C₈H₈N₂·ClH
Molecular Weight: 168.623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800384
CAS-Nummer: 57778-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR