Ethyl 2,2-Dimethyl-4-oxochroman-7-carboxylate

Lieferant: Apollo Scientific

OR471158-1G OR471158-250MG OR471158-5G
APOSOR471158-1GEA 713 EUR
APOSOR471158-1G APOSOR471158-250MG APOSOR471158-5G
Ethyl 2,2-Dimethyl-4-oxochroman-7-carboxylate
Ethyl 2,2-Dimethyl-4-oxochroman-7-carboxylate
Formel: C₁₄H₁₆O₄
Molecular Weight: 248,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036718
CAS-Nummer: 191611-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR