5-Chloro-4-methylbenzimidazole

Lieferant: Apollo Scientific

OR471274-1G OR471274-250MG OR471274-5G
APOSOR471274-1GEA 804 EUR
APOSOR471274-1G APOSOR471274-250MG APOSOR471274-5G
5-Chloro-4-methylbenzimidazole
5-Chloro-4-methylbenzimidazole
Formel: C₈H₇ClN₂
Molecular Weight: 166,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22557993
CAS-Nummer: 1360938-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR